H-Glu(OBzl)-OtBu·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-5-Benzyl 1-tert-butyl 2-aminopentanedioate hydrochloride, H-Glu(OBzl)-OtBu·HCl

4002216.0001 4002216.0005
E-3535.0001BAEA 180.91 CAD
E-3535.0001BA E-3535.0005BA
H-Glu(OBzl)-OtBu·HCl
H-Glu(OBzl)-OtBu·HCl

Sequence: H-Glu(OBzl)-OtBu · HCl


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₆H₂₄ClNO₄
MW: 329.821 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038897
CAS Number: 105590-97-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR