(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride

Supplier: BACHEM AMERICAS INC

4019074.0001 4019074.0005 4019074.0025
E-3115.0001BAEA 185.46 CAD
E-3115.0001BA E-3115.0005BA E-3115.0025BA
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride

Sequence: H-Asp(OMe)-OH · HCl


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₅H₉NO₄·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00038972
CAS Number: 16856-13-6
EINECS: 240-880-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR