H-Lys(Alloc)-OH

Supplier: BACHEM AMERICAS INC

4018971.0001 4018971.0005 4018971.0025
E-3065.0001BAEA 309.66 CAD
E-3065.0001BA E-3065.0005BA E-3065.0025BA
H-Lys(Alloc)-OH
H-Lys(Alloc)-OH

Sequence: H-Lys(Aloc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₀H₁₈N₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00237142
CAS Number: 6298-03-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR