H-His(1-Me)-OMe

Supplier: BACHEM AMERICAS INC

Synonyms: N'-Methyl-L-histidine methyl ester

4014917.0001 4014917.0005
E-2795.0001BAEA 748.67 CAD
E-2795.0001BA E-2795.0005BA
H-His(1-Me)-OMe
H-His(1-Me)-OMe

Sequence: H-His(1-Me)-OMe


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): H-His(τ-Me)-OMe

Formula: C₈H₁₃N₃O₂
MW: 183.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237978
CAS Number: 57519-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR