H-Cys(BZL)-OET·HCl

Supplier: BACHEM AMERICAS INC

4012395.0005 4012395.0025 4012395.0100
E-2720.0005BAEA 180.91 CAD
E-2720.0005BA E-2720.0025BA E-2720.0100BA
H-Cys(BZL)-OET·HCl
H-Cys(BZL)-OET·HCl

Sequence: H-Cys(Bzl)-OEt · HCl

Formula: C₁₂H₁₈ClNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00038909
CAS Number: 52844-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR