L-Valine methyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Val-OMe.HCl, L-2-Aminoisovaleric Acid Methyl Ester Hydrochloride, L-Methyl valinate hydrochloride, H-Val-OMe·HCl

4002561.0025 4002561.0100
E-2595.0025BAEA 180.91 CAD
E-2595.0025BA E-2595.0100BA
L-Valine methyl ester hydrochloride
L-Valine methyl ester hydrochloride

Sequence: H-Val-OMe · HCl


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.

Formula: C₆H₁₄ClNO₂
MW: 167.63 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012497
CAS Number: 6306-52-1
EINECS: 228-620-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR