L-Valine-tert-butyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

4000213.0005 4000213.0025
E-2590.0005BAEA 209 CAD
E-2590.0005BA E-2590.0025BA
L-Valine-tert-butyl ester hydrochloride
L-Valine-tert-butyl ester hydrochloride

Sequence: H-Val-OtBu · HCl


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.

Formula: C₉H₁₉NO₂·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00034863
CAS Number: 13518-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR