L(-)-Tryptophan

Supplier: BACHEM AMERICAS INC

4031215.0100 4031215.0250 4031215.1000
E-2500.0100BAEA 175.66 CAD
E-2500.0100BA E-2500.0250BA E-2500.1000BA
L(-)-Tryptophan
L(-)-Tryptophan

Sequence: H-Trp-OH

Formula: C₁₁H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00064340
CAS Number: 73-22-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR