(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride

Supplier: BACHEM AMERICAS INC

4000658.0025 4000658.0100
E-2270.0025BAEA 180.91 CAD
E-2270.0025BA E-2270.0100BA
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride

Sequence: H-Phe-OMe · HCl


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.

Formula: C₁₀H₁₄ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00012489
CAS Number: 7524-50-7
EINECS: 231-383-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR