N-Acetylornithine

Supplier: BACHEM AMERICAS INC

4002042.0001 4002042.0005
E-1150.0001BAEA 206.51 CAD
E-1150.0001BA E-1150.0005BA
N-Acetylornithine
N-Acetylornithine

Sequence: Ac-Orn-OH

Formula: C₇H₁₄N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00065115
CAS Number: 6205-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR