Fmoc-Lys(biotinyl)-Wang resin 0.3-0.6 mmol/g 200-400 mesh

Supplier: BACHEM AMERICAS INC

4037528.0001 4037528.0005
D-2705.0001BAEA 1497.37 CAD
D-2705.0001BA D-2705.0005BA
Fmoc-Lys(biotinyl)-Wang resin 0.3-0.6 mmol/g 200-400 mesh
Fmoc-Lys(biotinyl)-Wang resin

Sequence: Fmoc-Lys(biotinyl)-Wang resin (200-400 mesh)
Synonym(s): Fmoc-Lys(biotinyl)-p-alkoxybenzyl alcohol resin

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR