Anti-SLC51A Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
PA526837
CAPIPA526837EA 3335.52 CAD
CAPIPA526837
Anti-SLC51A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: SLC51A (solute carrier family 51, alpha subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR