Anti-A1BG Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
PA526454
CAPIPA526454EA 2894.45 CAD
CAPIPA526454
Anti-A1BG Rabbit Polyclonal Antibody
Antibodies
A1BG is a plasma glycoprotein of unknown function. The protein shows sequence similarity to the variable regions of some immunoglobulin supergene family member proteins.

Western Blot: 1:1000

Type: Primary
Antigen: A1BG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR