Anti-p19 Rabbit Polyclonal Antibody
Supplier: Thermo Scientific
PA526413
CAPIPA526413EA 3378.53 CAD
CAPIPA526413
Anti-p19 Rabbit Polyclonal Antibody
Antibodies
p19 is a member of the INK4 family of cyclin-dependent kinase inhibitors. This protein has been shown to form a stable complex with CDK4 or CDK6, and prevent the activation of the CDK kinases, thus function as a cell growth regulator that controls cell cycle G1 progression. The abundance of the transcript of this gene was found to oscillate in a cell-cycle dependent manner with the lowest expression at mid G1 and a maximal expression during S phase. The negative regulation of the cell cycle involved in this protein was shown to participate in repressing neuronal proliferation, as well asMore Product Information