Anti-GALNT3 Rabbit Polyclonal Antibody
Supplier: Thermo Scientific
PA526217
CAPIPA526217EA 3064.09 CAD
CAPIPA526217
Anti-GALNT3 Rabbit Polyclonal Antibody
Antibodies
GALNT3 encodes UDP-GalNAc transferase 3, a member of the GalNAc-transferases family. This family transfers an N-acetyl galactosamine to the hydroxyl group of a serine or threonine residue in the first step of O-linked oligosaccharide biosynthesis. Individual GalNAc-transferases have distinct activities and initiation of O-glycosylation is regulated by a repertoire of GalNAc-transferases. The protein is highly homologous to other family members, however the enzymes have different substrate specificities.
Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50, FlowMore Product Information