Anti-PDHX Rabbit Polyclonal Antibody
Supplier: Thermo Scientific
PA526170
CAPIPA526170EA 3335.52 CAD
CAPIPA526170
Anti-PDHX Rabbit Polyclonal Antibody
Antibodies
Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.
Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50