Anti-AMPD2 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
PA526127
CAPIPA526127EA 3327.27 CAD
CAPIPA526127
Anti-AMPD2 Rabbit Polyclonal Antibody
Antibodies
AMP deaminase plays a critical role in energy metabolism.

Flow Cytometry: 1:10-1:50, Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: AMPD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR