Anti-NKX2-1 Rabbit Polyclonal Antibody
Supplier: Thermo Scientific
PA525940
CAPIPA525940EA 3258.01 CAD
CAPIPA525940
Anti-NKX2-1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. This gene encodes a protein initially identified as a thyroid-specific transcription factor. The encoded protein binds to thyroglobulin promoter and regulates the expression of thyroid-specific genes but has also been shown to regulate the expression of genes involved in morphogenesis. Mutations and deletions in this gene are associated with benign hereditary chorea, choreoathetosis, congenital hypothyroidism, and neonatal respiratory distress, and may be associated with thyroid cancer. Multiple transcriptMore Product Information