Anti-ADCY10 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
PA525756
CAPIPA525756EA 3188.23 CAD
CAPIPA525756
Anti-ADCY10 Rabbit Polyclonal Antibody
Antibodies
Flow Cytometry: 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: ADCY10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR