Anti-BTG2 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
PA523630
CAPIPA523630EA 2895.26 CAD
CAPIPA523630
Anti-BTG2 Rabbit Polyclonal Antibody
Antibodies
The protein encoded by this gene is a member of the BTG/Tob family. This family has structurally related proteins that appear to have antiproliferative properties. This encoded protein is involved in the regulation of the G1/S transition of the cell cycle.

Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: BTG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR