Anti-GRIA4 Goat Polyclonal Antibody
Supplier: Thermo Scientific
PA5-18931
CAPIPA5-18931EA 3515.11 CAD
CAPIPA5-18931
Anti-GRIA4 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes composed of multiple subunits, arranged to form ligand-gated ion channels. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. The subunit encoded by this gene belongs to a family of AMPA -sensitive glutamate receptors, and isMore Product Information