Anti-MARCH10 Goat Polyclonal Antibody

Supplier: Thermo Scientific
PA5-18853
CAPIPA5-18853EA 3269.49 CAD
CAPIPA5-18853
Anti-MARCH10 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine and human based on sequence homology. MARCH10 is a member of the MARCH family of membrane-bound E3 ubiquitin ligases to target lysines in substrate proteins, thereby signaling their vesicular transport between membrane compartments .

Western Blot: 0.3-1 ug/ml

Type: Primary
Antigen: MARCH10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR