Anti-CADM4 Goat Polyclonal Antibody

Supplier: Thermo Scientific
PA5-18838
CAPIPA5-18838EA 3269.49 CAD
CAPIPA5-18838
Anti-CADM4 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, mouse and rat based on sequence homology.

Western Blot: 0.03-0.1 ug/ml

Type: Primary
Antigen: CADM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR