Anti-F13A1 Goat Polyclonal Antibody

Supplier: Thermo Scientific
PA5-18806
CAPIPA5-18806EA 3269.49 CAD
CAPIPA5-18806
Anti-F13A1 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse based on sequence homology. This gene encodes the coagulation factor XIII A subunit. Coagulation factor XIII is the last zymogen to become activated in the blood coagulation cascade. Plasma factor XIII is a heterotetramer composed of 2 A subunits and 2 B subunits. The A subunits have catalytic function, and the B subunits do not have enzymatic activity and may serve as plasma carrier molecules. Platelet factor XIII is comprised only of 2 A subunits, which are identical to those of plasma origin. Upon cleavage of the activation peptide byMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol F13A1
Environmentally Preferable
Antigen name Coagulation Factor 13 A1
Antigen synonyms coagulation factor XIII A chain|A1 polypeptide)|protein-glutamine gamma-glutamyltransferase A chain|A subunit|coagulation factor XIII|coagulation factor XIIIa|fibrinoligase|FSF|bA525O21.1 (coagulation factor XIII|factor XIIIa|TGase|A polypeptide|fibrin stabilizing factor|transglutaminase. plasma|transglutaminase A chain
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Goat
Isotype IgG
Western blot Yes
Cross adsorption No
Storage buffer TBS, pH 7.3, with 0.5% BSA and 0.02% Sodium azide
Storage temperature –20 °C, avoid freeze/thaw cycles

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR