Anti-B3GNT2 Goat Polyclonal Antibody

Supplier: Thermo Scientific
PA5-18803
CAPIPA5-18803EA 2904.29 CAD
CAPIPA5-18803
Anti-B3GNT2 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. This gene encodes a member of the beta-1,3-N-acetylglucosaminyltransferase family. This enzyme is a type II transmembrane protein. It prefers the substrate of lacto-N-neotetraose, and is involved in the biosynthesis of poly-N-acetyllactosamine chains.

Western Blot: 0.1-0.3 ug/ml

Type: Primary
Antigen: B3GNT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR