Anti-TREM2 Goat Polyclonal Antibody

Supplier: Thermo Scientific
PA5-18764
CAPIPA5-18764EA 3389.15 CAD
CAPIPA5-18764
Anti-TREM2 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with rat based on sequence homology. The protein encoded by this gene is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy.

Recommended Dilutions: Western Blot: 0.3-1 ug/ml

Type: Primary
Antigen: TREM2 (triggering receptor expressed on myeloid cells 2)
Clonality: polyclonal
Clone:
Conjugation:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR