Anti-Fumarase/FH Goat Polyclonal Antibody
Supplier: Thermo Scientific
PA5-18745
CAPIPA5-18745EA 3269.49 CAD
CAPIPA5-18745
Anti-Fumarase/FH Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, mouse and rat based on sequence homology. The protein encoded by this gene is an enzymatic component of the tricarboxylic acid cycle, or Krebs cycle, and catalyzes the formation of L-malate from fumarate. It exists in both a cytosolic form and an N-terminal extended form, differing only in the translation start site used. The N-terminal extended form is targeted to the mitochondrion, where the removal of the extension generates the same form as in the cytoplasm. It is similar to some thermostable class II fumarases and functions as aMore Product Information