Anti-GAB2 Goat Polyclonal Antibody
Supplier: Thermo Scientific
PA5-18705
CAPIPA5-18705EA 2904.29 CAD
CAPIPA5-18705
Anti-GAB2 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This gene is a member of the GRB2-associated binding protein gene family. These proteins contain pleckstrin homology domain, and bind SHP2 tyrosine phosphatase and GRB2 adapter protein. They act as adapters for transmitting various signals in response to stimuli through cytokine and growth factor receptors, and T- and B-cell antigen receptors. The protein encoded by this gene is the principal activator of phosphatidylinositol-3 kinase in response to activation of the high affinity IgE receptor. TwoMore Product Information