Anti-GPX1 Goat Polyclonal Antibody
Supplier: Thermo Scientific
PA5-18532
CAPIPA5-18532EA 3269.49 CAD
CAPIPA5-18532
Anti-GPX1 Goat Polyclonal Antibody
Antibodies
This gene encodes a member of the glutathione peroxidase family. Glutathione peroxidase functions in the detoxification of hydrogen peroxide, and is one of the most important antioxidant enzymes in humans. This protein is one of only a few proteins known in higher vertebrates to contain selenocysteine, which occurs at the active site of glutathione peroxidase and is coded by UGA, that normally functions as a translation termination codon. In addition, this protein is characterized in a polyalanine sequence polymorphism in the N-terminal region, which includes three alleles with five, six orMore Product Information