Anti-ROBO1/DUTT1 Goat Polyclonal Antibody
Supplier: Thermo Scientific
PA5-18460
CAPIPA5-18460EA 3269.49 CAD
CAPIPA5-18460
Anti-ROBO1/DUTT1 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. Bilateral symmetric nervous systems have special midline structures that establish a partition between the two mirror image halves. Some axons project toward and across the midline in response to long-range chemoattractants emanating from the midline. The product of this gene is a member of the immunoglobulin gene superfamily and encodes an integral membrane protein that functions in axon guidance and neuronal precursor cell migration. This receptor is activated by SLIT-family proteins,More Product Information