Anti-MAOA Goat Polyclonal Antibody

Supplier: Thermo Scientific
PA5-18420
CAPIPA5-18420EA 3269.49 CAD
CAPIPA5-18420
Anti-MAOA Goat Polyclonal Antibody
Antibodies
This gene encodes monoamine oxidase A, an enzyme that degrades amine neurotransmitters, such as dopamine, norepinephrine, and serotonin. The protein localizes to the mitochondrial outer membrane. The gene is adjacent to a related gene on the opposite strand of chromosome X. Mutation in this gene results in monoamine oxidase deficiency, or Brunner syndrome.

Recommended Dilutions: Western Blot: 0.1-0.3 ug/ml

Type: Primary
Antigen: MAOA (monoamine oxidase A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR