Anti-ABCE1 Goat Polyclonal Antibody
Supplier: Thermo Scientific
PA5-18412
CAPIPA5-18412EA 3268.55 CAD
CAPIPA5-18412
Anti-ABCE1 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. The protein encoded by this gene is a member of the superfamily of ATP-binding cassette transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies . This protein is a member of the OABP subfamily. Alternatively referred to as the RNase L inhibitor, this protein functions to block the activity of ribonuclease L. Activation of ribonuclease L leads to inhibition of protein synthesis in the 2-5A/RNase LMore Product Information