Anti-COL7A1 Goat Polyclonal Antibody
Supplier: Thermo Scientific
PA5-18390
CAPIPA5-18390EA 3269.49 CAD
CAPIPA5-18390
Anti-COL7A1 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, human, mouse and porcine based on sequence homology. This gene encodes the alpha chain of type VII collagen. The type VII collagen fibril, composed of three identical alpha collagen chains, is restricted to the basement zone beneath stratified squamous epithelia. It functions as an anchoring fibril between the external epithelia and the underlying stroma. Mutations in this gene are associated with all forms of dystrophic epidermolysis bullosa. In the absence of mutations, however, an acquired form of this disease can result from an autoimmuneMore Product Information