Anti-Nogo 66 receptor Goat Polyclonal Antibody

Supplier: Thermo Scientific
PA5-18380
CAPIPA5-18380EA 3517.33 CAD
CAPIPA5-18380
Anti-Nogo 66 receptor Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, mouse, porcine and rat based on sequence homology. This gene encodes the receptor for reticulon 4, oligodendrocyte myelin glycoprotein and myelin-associated glycoprotein. This receptor mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult central nervous system.

Recommended Dilutions: Western Blot: 0.3-1 ug/ml

Type: Primary
Antigen: Nogo 66 receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR