Anti-CCDC50 Goat Polyclonal Antibody
Supplier: Thermo Scientific
PA5-18320
CAPIPA5-18320EA 3269.49 CAD
CAPIPA5-18320
Anti-CCDC50 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, canine and rat based on sequence homology. This gene encodes a soluble, cytoplasmic, tyrosine-phosphorylated protein with multiple ubiquitin-interacting domains. Mutations in this gene cause nonsyndromic, postlingual, progressive sensorineural DFNA44 hearing loss. In mouse, the protein is expressed in the inner ear during development and postnatal maturation and associates with microtubule-based structures. This protein may also function as a negative regulator of NF-kB signaling and as an effector of epidermal growth factor -mediated cellMore Product Information