Anti-VPS45 Goat Polyclonal Antibody
Supplier: Thermo Scientific
PA5-18215
CAPIPA5-18215EA 3269.49 CAD
CAPIPA5-18215
Anti-VPS45 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, canine and mouse based on sequence homology. Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting genes involved in vesicle transport to vacuoles. This gene is a member of the Sec1 domain family, and shows a high degree of sequence similarity to mouse, rat and yeast Vps45. The exact function of this gene is not known, but its high expression in peripheral blood mononuclear cells suggests a roleMore Product Information