Anti-ZBTB32 Goat Polyclonal Antibody

Supplier: Thermo Scientific
PA5-18204
CAPIPA5-18204EA 3416.43 CAD
CAPIPA5-18204
Anti-ZBTB32 Goat Polyclonal Antibody
Antibodies
DNA-binding protein that binds to the to a 5'-TGTACAGTGT-3' core sequence. May function as a transcriptional transactivator and transcriptional repressor. Probably exerts its repressor effect by preventing GATA3 from binding to DNA. May play a role in regulating the differentiation and activation of helper T cells.

Recommended Dilutions: Western Blot: 2-5 ug/ml

Type: Primary
Antigen: ZBTB32 (zinc finger and BTB domain containing 32)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR