Anti-EPS8 Goat Polyclonal Antibody
Supplier: Thermo Scientific
PA5-18196
CAPIPA5-18196EA 3268.55 CAD
CAPIPA5-18196
Anti-EPS8 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with canine, human, mouse, porcine and rat based on sequence homology. This gene encodes a member of the EPS8 family. This protein contains one PH domain and one SH3 domain. It functions as part of the EGFR pathway, though its exact role has not been determined. Highly similar proteins in other organisms are involved in the transduction of signals from Ras to Rac and growth factor-mediated actin remodeling. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
Recommended Dilutions:More Product Information