Anti-GCIP-interacting protein p29 Goat Polyclonal Antibody

Supplier: Thermo Scientific
PA5-18108
CAPIPA5-18108EA 3269.49 CAD
CAPIPA5-18108
Anti-GCIP-interacting protein p29 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, canine and rat based on sequence homology. This gene encodes a nuclear protein that interacts with cyclin D-type binding-protein 1, which is thought to be a cell cycle regulator at the G1/S transition. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Recommended Dilutions: Western Blot: 0.5-3 ug/ml

Type: Primary
Antigen: GCIP-interacting protein p29
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR