Anti-A1AT Rabbit Polyclonal Antibody
Supplier: Thermo Scientific
PA5-16661
CAPIPA5-16661EA 2648.89 CAD
CAPIPA5-16661
Anti-A1AT Rabbit Polyclonal Antibody
Antibodies
PA5-16661 targets Alpha-1-Antitrypsin in IHC (P) applications and shows reactivity with Equine, Human, Mink, monkey, and baboon samples. Does not react with Canine, Chicken, Feline, Guinea Pig, Goat, Marsupial, mouse, Ovine, and Porcine samples The PA5-16661 immunogen is purified human serum alpha-1-AT. alpha-1-antitrypsin (alpha-1-AT) which exists in a number of genetic variants. MM variant is the most common. Alpha-1-AT is synthesized in the liver and it acts as an inhibitor of proteases such as trypsin, elastase, chymotrypsin, collagenase, leucocytic proteases, plasmin, and thrombin,More Product Information