Anti-LIM Kinase 1 Rabbit Polyclonal Antibody
Supplier: Thermo Scientific
PA514938
CAPIPA5-14938EA 3692.03 CAD
CAPIPA5-14938
Anti-LIM Kinase 1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. LIMK1, a member of the Ser/Thr protein kinase family, may be a component of an intracellular signaling pathway and may be involved in brain development. It phosphorylates and inactivates the actin binding/depolymerizing factor cofilin and induces actin cytoskeletal changes. The LIM domain interacts with the cytoplasmic domain of NRG1, and this cytoplasmic protein also binds ROCK1, whic phosphorylates LIMK1 on serine and/or threonine residues. Highest expression occurs in both adult and fetal nervous systems.More Product Information