Anti-ACVRL1 Rabbit Polyclonal Antibody
Supplier: Thermo Scientific
PA514921
CAPIPA5-14921EA 3335.52 CAD
CAPIPA5-14921
Anti-ACVRL1 Rabbit Polyclonal Antibody
Antibodies
ACVRL1 is a type I cell-surface receptor for the TGF-beta superfamily of ligands. It shares with other type I receptors a high degree of similarity in serine-threonine kinase subdomains, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. This protein, sometimes termed ALK1, shares similar domain structures with other closely related ALK or activin receptor-like kinase proteins that form a subfamily of receptor serine/threonine kinases. Mutations in this gene are associated with hemorrhagic telangiectasia type 2, also known asMore Product Information