Anti-HRAS Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
PA514863
CAPIPA5-14863EA 3406.75 CAD
CAPIPA5-14863
Anti-HRAS Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. HRAS belongs to the Ras oncogene family, whose members are related to the transforming genes of mammalian sarcoma retroviruses. These proteins function in signal transduction pathways. They can bind GTP and GDP, and they have intrinsic GTPase activity. HRAS undergoes a continuous cycle of de- and re-palmitoylation, which regulates its rapid exchange between the plasma membrane and the Golgi apparatus. Mutations in this gene cause Costello syndrome, a disease characterized by increased growth at the prenatalMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol HRAS
Environmentally Preferable
Antigen name HRas proto-oncogene, GTPase
Antigen synonyms C-H-RAS|CTLO|p21ras|HRAS1|RASH1|C-HA-RAS1|H-RASIDX|HAMSV|C-BAS/HAS
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Gel shift Yes
Flow cytometry Yes
ChIP Yes
Neutralisation Yes
Immunogen KLH conjugated synthetic peptide between 104-128 amino acids from the central region of human HRAS
Form Liquid
Molecular weight 21.3 kDa
Purification Ammonium sulfate precipitation, Size-exclusion - Dialysis
Concentration Lot-specific
Cross adsorption No
Storage buffer PBS with 0.09% Sodium azide
Storage temperature Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at –20 °C
Tested applications Immunohistochemistry

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR