Anti-CABP1 Rabbit Polyclonal Antibody
Supplier: Thermo Scientific
PA514540
CAPIPA5-14540EA 3258.01 CAD
CAPIPA5-14540
Anti-CABP1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine and rat based on sequence homology. CABP1 belongs to a subfamily of calcium binding proteins, which share similarity to calmodulin. Calcium binding proteins are an important component of calcium mediated cellular signal transduction. Expression of this protein was only detected in retina and brain. Study of the mouse homolog demonstrated that groups of cells expressing this protein are located in the center or inner border of the inner unclear layer of retina.
Immunohistochemistry: 1:10-1:50, Western Blot: 1:1000
Type: Primary
Antigen:More Product Information