Anti-PRKD1 Rabbit Polyclonal Antibody
Supplier: Thermo Scientific
PA513749
CAPIPA5-13749EA 3187.19 CAD
CAPIPA5-13749
Anti-PRKD1 Rabbit Polyclonal Antibody
Antibodies
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play distinct roles in cells. PKC mu is one of the PKC family members. The protein is a cytosolic serine-threonine kinase that binds to theMore Product Information