Anti-GLA Rabbit Polyclonal Antibody
Supplier: Thermo Scientific
PA513687
CAPIPA5-13687EA 3378.53 CAD
CAPIPA5-13687
Anti-GLA Rabbit Polyclonal Antibody
Antibodies
GLA is a homodimeric glycoprotein that hydrolyses the terminal alpha-galactosyl moieties from glycolipids and glycoproteins. This enzyme predominantly hydrolyzes ceramide trihexoside, and it can catalyze the hydrolysis of melibiose into galactose and glucose. A variety of mutations in this gene affect the synthesis, processing, and stability of this enzyme, which causes Fabry disease, a rare lysosomal storage disorder that results from a failure to catabolize alpha-D-galactosyl glycolipid moieties.
Flow Cytometry: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunofluorescence: 1:10-1:50,More Product Information