Anti-TNFAIP2 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
PA513542
CAPIPA5-13542EA 2895.26 CAD
CAPIPA5-13542
Anti-TNFAIP2 Rabbit Polyclonal Antibody
Antibodies
TNFAIP2 expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. The expression of TNFAIP2 was shown to be induced by retinoic acid in a cell line expressing a oncogenic version of the retinoic acid receptor alpha fusion protein, which suggested that its gene may be a retinoic acid target gene in acute promyelocytic leukemia.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:50 - 1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: TNFAIP2 (tumor necrosis factor, alpha-induced protein 2)
Clonality: polyclonal
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR