Anti-YBX1 Rabbit Polyclonal Antibody
Supplier: Thermo Scientific
PA513307
CAPIPA5-13307EA 3258.01 CAD
CAPIPA5-13307
Anti-YBX1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, mouse, rat and Xenopus based on sequence homology. YBX1 binds to splice sites in pre-mRNA and regulates splice site selection. This protein binds and stabilizes cytoplasmic mRNA and contributes to the regulation of translation by modulating the interaction between the mRNA and eukaryotic initiation factors. It binds to promoters that contain a Y-box (5'-CTGATTGGCCAA-3'), such as HLA class II genes. It regulates the transcription of numerous genes and promotes separation of DNA strands that contain mismatches or are modified by cisplatin. ItMore Product Information