Anti-RNF40 Rabbit Polyclonal Antibody
Supplier: Thermo Scientific
MA1014D650
CAPIMA1014D650EA 2208.41 CAD
CAPIMA1014D650
Anti-RNF40 Rabbit Polyclonal Antibody
Antibodies
For Western Blot, this antibody has non-specific bands at 65 kDa. The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein was reported to interact with the tumor suppressor protein RB1. Studies of the rat counterpart suggested that this protein may function as an E3 ubiquitin-protein ligase, and facilitate the ubiquitination and degradation of syntaxin 1, which is an essential component of the neurotransmitter release machinery.
Recommended Dilutions: Western Blot: 1 µg/ml, Immunofluorescence: 1:200More Product Information