Pierce™ NHS-Activated Agarose, Thermo Scientific
Supplier: Thermo Scientific
26196 26197 26198 26199 26200
CAPI26196EA 322.76 CAD
CAPI26196 CAPI26197 CAPI26198 CAPI26199 CAPI26200
Pierce™ NHS-Activated Agarose, Thermo Scientific
Electrophoresis Gels Agarose and Acrylamide
Thermo Scientific Pierce NHS-Activated Agarose is a high-quality, amine-reactive, beaded-agarose resin for rapid and stable immobilization of proteins, peptides and other ligands via primary amines.
  • Immobilise in a simple one-step reaction
  • No hazardous chemicals needed
  • Available as dry powder, spin columns or slurry
  • Leak-resistant chemistry makes it resuable
  • Coupling capacity >25 mg/ml for dry resin and >30 mg/ml for slurry
  • Affinity resin is adaptable to column and batch affinity chromatography techniques and FPLC applications
NHS-activatedMore Product Information